Learn More
Abnova™ Human VPS29 Partial ORF (NP_476528, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene belongs to a group of vacuolar protein sorting (VPS) genes that, when functionally impaired, disrupt the efficient delivery of vacuolar hydrolases. The protein encoded by this gene is a component of a large multimeric complex, termed the retromer complex, which is involved in retrograde transport of proteins from endosomes to the trans-Golgi network. This VPS protein may be involved in the formation of the inner shell of the retromer coat for retrograde vesicles leaving the prevacuolar compartment. Alternative splice variants encoding different isoforms, and usage of multiple polyadenylation sites have been found for this gene. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_476528 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51699 |
Molecular Weight (g/mol) | 36.63kDa |
Name | VPS29 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | MAGHRLVLVLGDLHIPHRCNSLPAKFKKLLVPGKIQHILCTGNLCTKESYDYLKTLAGDVHIVRGDFDENLNYPEQKVVTVGQFKIGLIHGHQVIPWGD |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.