Learn More
Abnova™ Human ZNF197 Partial ORF (NP_001020026.1, 161 a.a. - 267 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010168-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3' UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. This gene is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: IAAEICPHPPTDLVAFNLQDPQHDSPAPEASALSQEENPRNQLMALMLLTAQPQELVMFEEVSVCFTSEEWACLGPIQRALYWDVMLENYGNVTSLGYRKYRRQRNKSpecifications
NP_001020026.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IAAEICPHPPTDLVAFNLQDPQHDSPAPEASALSQEENPRNQLMALMLLTAQPQELVMFEEVSVCFTSEEWACLGPIQRALYWDVMLENYGNVTSLGYRKYRRQRNK | |
RUO | |
ZNF197 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10168 | |
ZNF197 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D3S1363E/P18/VHLaK/ZKSCAN9/ZNF166/ZNF20 | |
ZNF197 | |
Recombinant | |
wheat germ expression system |