Learn More
Abnova™ Human ZNF197 Partial ORF (NP_001020026.1, 161 a.a. - 267 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001020026.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10168 |
Molecular Weight (g/mol) | 37.51kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16152026
|
Abnova™
H00010168-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16162026
|
Abnova™
H00010168-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3' UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. This gene is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: IAAEICPHPPTDLVAFNLQDPQHDSPAPEASALSQEENPRNQLMALMLLTAQPQELVMFEEVSVCFTSEEWACLGPIQRALYWDVMLENYGNVTSLGYRKYRRQRNKSpecifications
NP_001020026.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D3S1363E/P18/VHLaK/ZKSCAN9/ZNF166/ZNF20 | |
ZNF197 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10168 | |
ZNF197 (Human) Recombinant Protein (Q01) | |
IAAEICPHPPTDLVAFNLQDPQHDSPAPEASALSQEENPRNQLMALMLLTAQPQELVMFEEVSVCFTSEEWACLGPIQRALYWDVMLENYGNVTSLGYRKYRRQRNK | |
RUO | |
ZNF197 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |