Learn More
Abnova™ Human ZNF213 Partial ORF (NP_004211, 133 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007760-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
C2H2 zinc finger proteins, such as ZNF213, have bipartite structures in which one domain binds DNA or RNA and the other modulates target gene expression.[supplied by OMIM]
Sequence: QDVPSEEAEPEAAGRGSQATGPPPTVGARRRPSVPQEQHSHSAQPPALLKEGRPGETTDTCFVSGVHGPVALGDIPFYFSREEWGTLDPAQRDLFWDIKRENSRNTTLGFSpecifications
NP_004211 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QDVPSEEAEPEAAGRGSQATGPPPTVGARRRPSVPQEQHSHSAQPPALLKEGRPGETTDTCFVSGVHGPVALGDIPFYFSREEWGTLDPAQRDLFWDIKRENSRNTTLGF | |
RUO | |
ZNF213 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7760 | |
ZNF213 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CR53/ZKSCAN21 | |
ZNF213 | |
Recombinant | |
wheat germ expression system |