missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF281 Partial ORF (NP_036614, 787 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Sequence: KLTSQKEQKNLESSTGFQIPSQELASQIDPQKDIEPRTTYQIENFAQAFGSQFKSGSRVPMTFITNSNGEVDHRVRTSVSDFSGYTNMMSDVSEPCSTRVKTPTSQS
Specifications
Specifications
Accession Number | NP_036614 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23528 |
Molecular Weight (g/mol) | 37.51kDa |
Name | ZNF281 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | KLTSQKEQKNLESSTGFQIPSQELASQIDPQKDIEPRTTYQIENFAQAFGSQFKSGSRVPMTFITNSNGEVDHRVRTSVSDFSGYTNMMSDVSEPCSTRVKTPTSQS |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Abnova™ Human ZNF281 Partial ORF (NP_036614, 787 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur