missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF550 Full-length ORF (AAH53858.1, 1 a.a. - 413 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4630.00 SEK - 7020.00 SEK
Specifications
Accession Number | AAH53858.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 162972 |
Molecular Weight (g/mol) | 73.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16118953
|
Abnova™
H00162972-P01.25ug |
25 ÎĽg |
7020.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16108953
|
Abnova™
H00162972-P01.10ug |
10 ÎĽg |
4630.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence: MLVTFKDVAVTFTREEWRQLDLAQRTLYREVMLETCGLLVSLGHRVPKPELVHLLEHGQELWIVKRGLSHATCAGDRAQVHTREPTTYPPVLSERAFLRGSLTLESSTSSDSRLGRARDEEGLLEMQKGKVTPETDLHKETHLGKVSLEGEGLGTDDGLHSRALQEWLSADVLHECDSQQPGKDALIHAGTNPYKCKQCGKGFNRKWYLVRHQRVHTGMKPYECNACGKAFSQSSTLIRHYLIHTGEKPYKCLECGKAFKRRSYLMQHHPIHTGEKPYECSQCRKAFTHRSTFIRHNRTHTGEKPFECKECEKAFSNRAHLIQHYIIHTGEKPYDCMACGKAFRCSSELIQHQRIHTGEKPYECTQCGKAFHRSTYLIQHSVIHTGEMPYKCIECGKAFKRRSHLLQHQRVHTSpecifications
AAH53858.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLVTFKDVAVTFTREEWRQLDLAQRTLYREVMLETCGLLVSLGHRVPKPELVHLLEHGQELWIVKRGLSHATCAGDRAQVHTREPTTYPPVLSERAFLRGSLTLESSTSSDSRLGRARDEEGLLEMQKGKVTPETDLHKETHLGKVSLEGEGLGTDDGLHSRALQEWLSADVLHECDSQQPGKDALIHAGTNPYKCKQCGKGFNRKWYLVRHQRVHTGMKPYECNACGKAFSQSSTLIRHYLIHTGEKPYKCLECGKAFKRRSYLMQHHPIHTGEKPYECSQCRKAFTHRSTFIRHNRTHTGEKPFECKECEKAFSNRAHLIQHYIIHTGEKPYDCMACGKAFRCSSELIQHQRIHTGEKPYECTQCGKAFHRSTYLIQHSVIHTGEMPYKCIECGKAFKRRSHLLQHQRVHT | |
RUO | |
ZNF550 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
162972 | |
ZNF550 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC41917 | |
ZNF550 | |
Recombinant | |
wheat germ expression system |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human ZNF550 Full-length ORF (AAH53858.1, 1 a.a. - 413 a.a.) Recombinant Protein with GST-tag at N-terminal