missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human ZRF1 Partial ORF (XP_168590.4, 408 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16115616

Abnova™ Human ZRF1 Partial ORF (XP_168590.4, 408 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16115616
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16115616

Brand: Abnova™ H00027000Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is a member of the M-phase phosphoprotein (MPP) family. The gene encodes a phosphoprotein with a J domain and a Myb DNA-binding domain which localizes to both the nucleus and the cytosol. The protein is capable of forming a heterodimeric complex that associates with ribosomes, acting as a molecular chaperone for nascent polypeptide chains as they exit the ribosome. This protein was identified as a leukemia-associated antigen and expression of the gene is upregulated in leukemic blasts. Also, chromosomal aberrations involving this gene are associated with primary head and neck squamous cell tumors. This gene has a pseudogene on chromosome 6. Alternatively spliced variants which encode different protein isoforms have been described. [provided by RefSeq]

Sequence: LDPHQKDDINKKAFDKFKKEHGVVPQADNATPSERFEGPYTDFTPWTTEEQKLLEQALKTYPVNTPERWEKIAEAVPGRTKKDCMKRYKELVEMVKAKKAA

Specifications

Accession Number XP_168590.4
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27000
Molecular Weight (g/mol) 36.85kDa
Name ZRF1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen LDPHQKDDINKKAFDKFKKEHGVVPQADNATPSERFEGPYTDFTPWTTEEQKLLEQALKTYPVNTPERWEKIAEAVPGRTKKDCMKRYKELVEMVKAKKAA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MPHOSPH11/MPP11/ZRF1/ZUO1
Common Name DNAJC2
Gene Symbol DNAJC2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human ZRF1 Partial ORF (XP_168590.4, 408 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.