missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human Thrombin (Coagulation Factor II) Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
26714.50 SEK - 91156.79 SEK
Specifications
Gene ID (Entrez) | 2147 |
---|---|
Molecular Weight (g/mol) | 36 kDa, glycosylated |
Species | Human |
Content And Storage | This protein is stable for 1 year from the date of receipt if stored between -20 to -80°C. Repeated freeze thaw cycles should be avoided. |
Expression System | Human embryonic kidney 293 (HEK293) human cell expression system |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881426
|
Proteintech™
HZ-3010-1000UG |
1000 ÎĽg |
26714.50 SEK
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16801436
|
Proteintech™
HZ-3010-5000UG |
5000 ÎĽg |
60703.40 SEK
5000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16891426
|
Proteintech™
HZ-3010-10000UG |
10 x 1000 ÎĽg | N/A | ||||||
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
2147 | |
Human | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 95% |
36 kDa, glycosylated | |
This protein is stable for 1 year from the date of receipt if stored between -20 to -80°C. Repeated freeze thaw cycles should be avoided. | |
Lyophilized |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech™ HumanKine™ Recombinant Human Thrombin (Coagulation Factor II) Protein