missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huntingtin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19957-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of HTT.
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.Specifications
Huntingtin | |
Polyclonal | |
Unconjugated | |
HTT | |
HD, HD protein, HTT, huntingtin, Huntington disease protein, IT15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15194, 3064 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42858, P42859 | |
Htt | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Huntingtin Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction