missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HVEM (CD270) Mouse anti-Human, Oligo, Clone: CW10 (also known as CW10), BD™ AbSeq ™
Mouse Monoclonal Antibody
Brand: BD Biosciences 940097
This item is not returnable.
View return policy
Description
HVEM (CD270) Monoclonal antibody specifically detects HVEM (CD270) in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
HVEM (CD270) | |
Monoclonal | |
Oligo | |
TNFRSF14; HVEM; HVEA; LIGHTR ; ATAR ; TR2 | |
Human Recombinant Protein | |
RUO | |
Human | |
IgG1 Îş |
Single Cell 3' Sequencing | |
Clone CW10 (also known as CW10) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HVEM (CD270) Mouse anti-Human, Oligo, Clone: CW10 (also known as CW10), BD™ AbSeq ™ > 25 Tests; Oligo
Spot an opportunity for improvement?Share a Content Correction