Learn More
HYAL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18139-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This polyclonal antibody is raised against 29-460 aa of human HYAL4. The apparent molecular weight is larger than the calculated molecular weight of 54 kDa, which is likely due to posttranslational modification, presumably glycosylation.
Endo-hyaluronidase that degrades hyaluro to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.Specifications
HYAL4 | |
Polyclonal | |
Unconjugated | |
HYAL4 | |
Hyal 4, HYAL4, Hyaluronidase 4, hyaluronoglucosaminidase 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23553, 404783, 77042 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q05A56, Q2M3T9 | |
Hyal4 | |
HYAL4 Fusion Protein Ag12766 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |