Läs mer
HYDRANAL™ - Methanol Rapid, Honeywell Fluka™
Produktkod. 17191296 Handla allt Honeywell Fluka Produkter
Change view
Kvantitet:
500 ml
Produktkod. Kvantitet
17191296 500 ml
1 options
Produktkod. 17191296 Leverantör Honeywell Fluka Leverantörsnummer 37817500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Medium for accelerated volumetric one-component Karl Fischer titration
Hydranal™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material HYDRANAL™ - Methanol Rapid
CAS 288-32-4, 67-56-1, 7446-09-5
Kvantitet 500 ml