Learn More
Hydrogen Peroxide, technical, 35 wt% solution in water, stabilized, Thermo Scientific Chemicals
Product Code. 13248003 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. Quantity Unit Size
13248003 5 L 5 L
1 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 13248003 Supplier Thermo Scientific Acros Supplier No. 444380050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Beräknad leverans: 25-11-2026
Add to basket
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7722-84-1, 7732-18-5
Boiling Point 108.0°C
Melting Point -33.0°C
Molecular Formula H2O2
Linear Formula H2O2
MDL Number MFCD00011333
Density 1.1350g/mL
Merck Index 15,4835
Synonym perhydrol
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether, insoluble in petroleum ether
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name hydrogen peroxide
Molecular Weight (g/mol) 34
PubChem CID 784
ChEBI CHEBI:16240
Formula Weight 34
Grade Technical
Packaging Plastic Drum
Specific Gravity 1.135
Quantity 5 L
Chemical Name or Material Hydrogen Peroxide, 35 wt% solution in water, stabilized