Learn More
Ansell HyFlex™ 11-730 Polyurethane-Coated Gloves with Intercept™ Technology
Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.
Brand: Ansell 11730110
2042.27 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- A combination of comfort, sure handling, and cut resistance
- Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
- Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
- Designed for use in dry as well as slightly oily environments
Specifications
EN 407:2004, CE 0493, EN 420:2003 + A1:2009 | |
Polyurethane | |
ANSI A5 | |
Yes | |
Cut- and Abrasion-resistant Glove | |
4X43D | |
Unisex | |
Rough | |
13 |
Knit | |
Gray | |
Specific | |
Intercept Technology Yarn | |
11 | |
Metalwork, Parts Assembly, and Press Injection Molding | |
Knit | |
11-730 |
Product Content Correction
Product Title
Ansell HyFlex™ 11-730 Polyurethane-Coated Gloves with Intercept™ Technology > 11
Spot an opportunity for improvement?Share a Content Correction