Learn More
IBA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IBA1 |
---|---|
Concentration | 420 ÎĽg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
The IBA1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IBA1. This antibody recognizes human, mouse, rat antigen. The IBA1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
IBA1 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
420 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
199 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.