Läs mer
Tocris Bioscience™ ICA 121431
GREENER_CHOICE
Produktkod. 30002050
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30002050 10 mg
30002058 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30002050 Leverantör Tocris Bioscience™ Leverantörsnummer 5066/10
Potent and selective NaV1.3 and NaV1.1 channel blocker
Potent and selective inhibitor of human NaV1.3 and NaV1.1 channels (IC50 values are 13 and 23 nM respectively). Exhibits up to 1,000 fold selectivity against other TTX-sensitive or resistant sodium channels (IC50 values are >10 μM for human NaV1.5 and NaV1.8 channels). Interacts with an inhibitory interaction site distinct from those bound by TTX and local anesthetic-like modulators.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 313254-51-2
Molekylformel C23H19N3O3S2
Specifikationer
Kemiskt namn eller material ICA 121431
Kvantitet 10 mg
Formel vikt 449.55
Fysisk form Solid
Färg White
Renhet >98%
Molekylvikt (g/mol) 449.543