Läs mer
ICAM-1/CD54 Antibody (MEM-111) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB500-318-25ug
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifikationer
| ICAM-1/CD54 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 0.5-4 μg/mL, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 10 μg/mL, Immunohistochemistry-Frozen, CyTOF-ready | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Mouse | |
| 110 kDa | |
| 25 μg | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology | |
| 3383 | |
| Human, Rat, Bovine | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MEM-111 | |
| Unconjugated | |
| P05362 | |
| ICAM1 | |
| Raji human Burkitt's lymphoma cell line (NM_000201.2). | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody MEM-111 reacts with an extracellular epitope CD54 (ICAM-1), a 85-110 kDa type I transmembrane glycoprotein (receptor for rhinovirus). The expression of CD54 is upregulated by activation; it is expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells. HLDA VI; WS Code AS A049 | |
| Store at 4°C. Do not freeze. | |
| IgG2a |