missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10831-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CD54, (MW 90,000) antigen is the Intercellular Adhesion Molecule-1 (ICAM-1) and is the ligand for LFA-1 mediated adhesion. CD54 is weakly expressed on peripheral blood leukocytes. Upon activation the antigen is expressed strongly on B cells, T cells, macrophages and granulocytes. ICAM-1 is also inducible on endothelial cells, epithelial cells and fibroblasts.Specifications
ICAM-1 | |
Polyclonal | |
Unconjugated | |
ICAM1 | |
BB2, CD54, ICAM 1, ICAM1, P3.58 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3383 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05362 | |
ICAM1 | |
ICAM-1 Fusion Protein Ag1188 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |