missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11166-2-AP-20UL
This item is not returnable.
View return policy
Description
IDI1 encodes a peroxisomally-localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP), which are the substrates for the successive reaction that results in the synthesis of farnesyl diphosphate and, ultimately, cholesterol. It has been shown in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy that there is reduction in IPP isomerase activity.Specifications
IDI1 | |
Polyclonal | |
Unconjugated | |
IDI1 | |
IDI1, IPP isomerase 1, IPP1, IPPI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
319554, 3422, 89784 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35760, P58044, Q13907 | |
Idi1 | |
IDI1 Fusion Protein Ag1636 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction