missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1706.43 SEK - 4863.32 SEK
Specifications
Antigen | IDO2 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Along with the enzymes encoded by the INDO and TDO2 genes, the enzyme encoded by the INDOL1 gene metabolizes tryptophan in the kynurenine pathway.Specifications
IDO2 | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q6ZQW0 | |
169355 | |
IDO2 Fusion Protein Ag21866 | |
Primary | |
-20°C | |
IDO2 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IDO 2, IDO2, INDOL1, indoleamine 2,3 dioxygenase 2 | |
IDO2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IDO2 Rabbit anti-Human, Polyclonal, Proteintech