Läs mer
IFN-gamma Mouse anti-Chicken, Clone: MT6C2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-18280-1000UG
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Chicken samples. It is validated for ELISA, ELISPOT
Specifikationer
| IFN-gamma | |
| Monoclonal | |
| 0.5 mg/ml | |
| 0.2 um filtered PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Chicken | |
| Purified |
| ELISA | |
| MT6C2 | |
| Unconjugated | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Recombinant chicken IFN-gamma | |
| 1000 μg | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| 3458 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |