missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNAR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1706.43 SEK - 4863.32 SEK
Specifications
Antigen | IFNAR1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon and Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.Specifications
IFNAR1 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
P17181 | |
3454 | |
IFNAR1 Fusion Protein Ag3739 | |
Primary | |
-20°C | |
IFNAR1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AVP, CRF2 1, IFN alpha REC, IFN alpha/beta receptor 1, IFN R 1, IFNAR, IFNAR1, IFNBR, IFRC, Type I interferon receptor 1 | |
IFNAR1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IFNAR1 Rabbit anti-Human, Polyclonal, Proteintech