missing translation for 'onlineSavingsMsg'
Learn More

IFNGR2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10266-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16836543

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal domit or X-linked inheritance.
Specifications

Specifications

IFNGR2
Polyclonal
Unconjugated
IFNGR2
AF 1, IFGR2, IFN gamma R2, IFN gamma receptor 2, IFNGR2, IFNGT1, Interferon gamma receptor 2, Interferon gamma transducer 1
Rabbit
Antigen Affinity Chromatography
RUO
3460
-20°C
Liquid
Immunoprecipitation, Flow Cytometry, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P38484
IFNGR2
IFNGR2 Fusion Protein Ag0317
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers