missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNGR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | IFNGR2 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunoprecipitation, Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836543
|
Proteintech
10266-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826543
|
Proteintech
10266-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal domit or X-linked inheritance.Specifications
IFNGR2 | |
Immunoprecipitation, Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P38484 | |
3460 | |
IFNGR2 Fusion Protein Ag0317 | |
Primary | |
-20°C | |
IFNGR2 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AF 1, IFGR2, IFN gamma R2, IFN gamma receptor 2, IFNGR2, IFNGT1, Interferon gamma receptor 2, Interferon gamma transducer 1 | |
IFNGR2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |