Learn More
IFT27 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15017-1-AP-20UL
This item is not returnable.
View return policy
Description
RABL4 belongs to the small GTPase superfamily, Ras family. RABL4 possesses GTPase activity By similarity. This gene encodes a putative GTP-binding protein similar to RAY/RAB1C. The protein is ras-related, but the function is unknown.Specifications
IFT27 | |
Polyclonal | |
Unconjugated | |
IFT27 | |
IFT27, Rab like protein 4, RABL4, RAYL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11020 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BW83 | |
IFT27 | |
IFT27 Fusion Protein Ag6967 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
IFT27 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction