missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT81 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IFT81 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899023
|
Proteintech
11744-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889023
|
Proteintech
11744-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.Specifications
IFT81 | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Canine | |
O35594, Q8WYA0 | |
100856715, 12589, 28981 | |
IFT81 Fusion Protein Ag2339 | |
Primary | |
-20°C | |
IFT81 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CDV 1, CDV 1R, CDV1, CDV1R, IFT81 | |
IFT81 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IFT81 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech