missing translation for 'onlineSavingsMsg'
Learn More

IFT88 Mouse anti-Human, Mouse, Rat, Clone: 4A4G5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60227-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877245

  • 3313.85 SEK / 150µL
Estimated Shipment: 18-05-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.

This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

IFT88
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13099, Q61371
IFT88
IFT88 Fusion Protein Ag4980
150 μL
Primary
Human, Mouse, Rat
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
4A4G5
Unconjugated
IFT88
IFT88, TPR repeat protein 10, TTC10
Mouse
Protein G
RUO
21821, 305918, 8100
Antibody
IgG1
Videos
SDS
Documents
Certificates
Special Offers

Special Offers