Learn More
IGFBP6 Mouse anti-Human, Unconjugated, Clone: 1D12B3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67567-1-IG-150UL
 View more versions of this product
Product Code. 16890266
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The IGFBP6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IGFBP6. This antibody recognizes human antigen. The IGFBP6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IGFBP6
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1D12B3
Unconjugated
150 μL
3489
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IGFBP6 Mouse anti-Human, Unconjugated, Clone: 1D12B3, Proteintech > 150 μL