Learn More
IGFBP6 Mouse anti-Human, Unconjugated, Clone: 1D12B3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67567-1-IG-150UL
 View more versions of this product
Product Code. 16890266
  • 4652.90 SEK / 150µL
Description
The IGFBP6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IGFBP6. This antibody recognizes human antigen. The IGFBP6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IGFBP6
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1D12B3
Unconjugated
150 μL
3489
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IGFBP6 Mouse anti-Human, Unconjugated, Clone: 1D12B3, Proteintech > 150 μL