Learn More
IgG Heavy Chain Mouse anti-Rabbit, Unconjugated, Clone: 1C9A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66467-1-IG-150UL
Product Code. 16808375
  • 2144.88 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The Heavy chain of Rabbit IgG antibody from Proteintech is a mouse monoclonal antibody generated with protein of rabbit Heavy chain of Rabbit IgG. This antibody recognizes rabbit antigen. The Heavy chain of Rabbit IgG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IgG Heavy Chain
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
IgG1
Western Blot, ELISA
1C9A5
Unconjugated
150 ÎĽL
Rabbit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IgG Heavy Chain Mouse anti-Rabbit, Unconjugated, Clone: 1C9A5, Proteintech > 150 ÎĽL