missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Mouse anti-Bovine, Unconjugated, Clone: 1C5C5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IgG |
---|---|
Clone | 1C5C5 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800346
|
Proteintech
67615-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890336
|
Proteintech
67615-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Bovine IgG antibody from Proteintech is a mouse monoclonal antibody generated with peptide of bovine Bovine IgG. This antibody recognizes bovine antigen. The Bovine IgG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
IgG | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
IgG2a |
1C5C5 | |
Western Blot, ELISA | |
Unconjugated | |
Bovine | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IgG Mouse anti-Bovine, Unconjugated, Clone: 1C5C5, Proteintech