Learn More
IgG1 Rat, PE, Clone: RTK2071, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60075PE.1
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17198891
  • 729.00 SEK / 25µg
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • The RTK2071 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
Specifications
IgG1
Monoclonal
PE
25 μg
IgG1 κ
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
RTK2071
Rat
Isotype Control (Primary)
Other