missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1 Rat, PE, Clone: RTK2071, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
729.00 SEK - 1763.10 SEK
Specifications
Antigen | IgG1 |
---|---|
Clone | RTK2071 |
Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17198891
|
Stemcell Technologies
60075PE.1 |
25 μg |
729.00 SEK
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17188891
|
Stemcell Technologies
60075PE |
100 μg |
1763.10 SEK
100µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
- The RTK2071 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
IgG1 | |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
PE | |
IgG1 κ |
RTK2071 | |
Monoclonal | |
Rat | |
Isotype Control (Primary) |