missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG3 Lambda Human, Alexa Fluor 532, Isotype Control, Novus Biologicals™
Human Polyclonal Isotype Control
Brand: Bio-Techne NBP3-06871AF532
This item is not returnable.
View return policy
Description
IgG3 Lambda Polyclonal antibody specifically detects IgG3 Lambda in Human samples. It is validated for ELISA, Control
Specifications
IgG3 Lambda | |
Polyclonal | |
50mM Sodium Borate | |
Human myeloma serum | |
RUO | |
Store at 4°C in the dark. | |
IgG3 λ |
ELISA | |
Alexa Fluor 532 | |
Human | |
0.1 mL | |
Human | |
Isotype Controls |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IgG3 Lambda Human, Alexa Fluor 532, Isotype Control, Novus Biologicals™ > 0.1 mL; Alexa Fluor 532
Spot an opportunity for improvement?Share a Content Correction