missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Lambda Human, Alexa Fluor 532, Isotype Control, Novus Biologicals™
Human Polyclonal Isotype Control
Brand: Bio-Techne NBP3-06875AF532
This item is not returnable.
View return policy
Description
IgM Lambda Polyclonal antibody specifically detects IgM Lambda in Human samples. It is validated for ELISA, Control
Specifications
IgM Lambda | |
Polyclonal | |
50mM Sodium Borate | |
Human myeloma serum | |
RUO | |
Human | |
Isotype Controls |
ELISA | |
Alexa Fluor 532 | |
Human | |
0.1 mL | |
3507 | |
Store at 4°C in the dark. | |
IgM λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IgM Lambda Human, Alexa Fluor 532, Isotype Control, Novus Biologicals™ > 0.1 mL; Alexa Fluor 532
Spot an opportunity for improvement?Share a Content Correction