Learn More
Rat IgM Isotype Control (eBRM), FITC, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Invitrogen 11-4341-82
Description
Description: The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control FITC has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgM is an immunoglobulin M isotype antibody produced by B cells in rats. It plays a crucial role in the immune system by recognizing and neutralizing antigens, such as bacteria and viruses. Rat IgM has been extensively studied and is widely used in research applications, including immunology, microbiology, and biotechnology. It is commonly used as a control in experiments, as well as a diagnostic tool for detecting infectious diseases and autoimmune disorders. Researchers have developed various techniques for producing and purifying Rat IgM, including hybridoma technology, protein A/G chromatography, and affinity chromatography. Rat IgM is a valuable tool in the field of immunology, and its use has contributed to a better understanding of the immune system and the development of new treatments for various diseases.
Specifications
Rat IgM | |
eBRM | |
FITC | |
Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.