missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKZF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IKZF2 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881824
|
Proteintech
13554-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871824
|
Proteintech
13554-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predomitly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.Specifications
IKZF2 | |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P81183, Q9UKS7 | |
22779, 22807, 301476 | |
IKZF2 Fusion Protein Ag4357 | |
Primary | |
-20°C | |
IKZF2 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HELIOS, IKZF2, Zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
Ikzf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKZF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech