missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKZF5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IKZF5 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883265
|
Proteintech
24780-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873265
|
Proteintech
24780-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA-binding protein that binds to the 5′GNNTGTNG-3′ core sequence. Transcriptional repressor.Specifications
IKZF5 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q9H5V7 | |
64376 | |
IKZF5 Fusion Protein Ag20405 | |
Primary | |
-20°C | |
IKZF5 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IKZF5, PEGASUS, Zinc finger protein Pegasus, ZNFN1A5 | |
IKZF5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKZF5 Rabbit anti-Human, Polyclonal, Proteintech