Learn More
IL-10R alpha Mouse anti-Human, Clone: 714212, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems MAB2742-SP
Encompass
Product Code. 15725574
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
IL-10R alpha Mouse anti-Human, Clone: 714212, R&D Systems™