missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-29 Mouse anti-Human, Clone: 1H3A3, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IL-29 |
---|---|
Clone | 1H3A3 |
Concentration | 1.21 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897785
|
Proteintech
66163-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887785
|
Proteintech
66163-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL29 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28A (IL28A), and interleukin 28B (IL28B) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).Specifications
IL-29 | |
1.21 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cytokine Zcyto21, IFN lambda 1, IFNL1, IL 29, IL29, Interferon lambda 1, Interleukin 29, ZCYTO21 | |
IFNL1 | |
IgG1 | |
Protein G | |
Antibody |
1H3A3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8IU54 | |
282618 | |
IL-29 Fusion Protein Ag12444 | |
Primary | |
-20°C | |
IFNL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL-29 Mouse anti-Human, Clone: 1H3A3, Proteintech