missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL36RN Mouse anti-Human, Clone: 5F11G6, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IL36RN |
---|---|
Clone | 5F11G6 |
Concentration | 1.37 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887375
|
Proteintech
60298-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877375
|
Proteintech
60298-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.Specifications
IL36RN | |
1.37 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FIL1, FIL1 delta, FIL1(DELTA), FIL1D, IL 1 delta, IL 1 related protein 3, IL 1F5, IL 1HY1, IL 1L1, IL 1ra homolog 1, IL 1RP3, IL1F5, IL1HY1, IL1L1, IL1RP3, Interleukin 1 delta, Interleukin 1 family member 5, Interleukin 1 HY1, Interleukin 1 like protein 1 | |
IL36RN | |
IgG2b | |
Protein A | |
Antibody |
5F11G6 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UBH0 | |
26525 | |
IL36RN Fusion Protein Ag3780 | |
Primary | |
-20°C | |
IL36RN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL36RN Mouse anti-Human, Clone: 5F11G6, Proteintech