missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL37 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IL37 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899213
|
Proteintech
11863-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889213
|
Proteintech
11863-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
IL37 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NZH6 | |
27178 | |
IL37 Fusion Protein Ag2441 | |
Primary | |
-20°C | |
IL37 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FIL1, FIL1 zeta, FIL1(ZETA), FIL1Z, IL 1 zeta, IL 1F7, IL 1H4, IL 1RP1, IL 1X, IL1F7, IL1H4, IL1RP1, Interleukin 1 family member 7, Interleukin 1 homolog 4, Interleukin 1 related protein, Interleukin 1 zeta | |
IL37 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL37 Rabbit anti-Human, Polyclonal, Proteintech