missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
IL6R Antibody (rhPM-1 (Tocilizumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-75193
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence, Inhibition Activity.
Specifikationer
| IL6R | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300 | |
| IL6R | |
| This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions. | |
| 0.2 mg | |
| 3570.0 | |
| Human | |
| Purified |
| ELISA, Immunofluorescence, Inhibition Assays | |
| rhPM-1 (Tocilizumab) | |
| ELISA 1:100 - 1:2000, Immunofluorescence, Inhibition Activity | |
| biosimilar, biosimilars, CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membra | |
| Rabbit | |
| Protein A purified | |
| Primary | |
| Tocilizumab binds specifically to both the membrane bound and soluble forms of human IL-6R. The IL6 receptor is a protein complex consisting of an IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. Interleukin 6 (IL6) is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering