missing translation for 'onlineSavingsMsg'
Learn More

ILVBL Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24738-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883225

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.
Specifications

Specifications

ILVBL
Polyclonal
Unconjugated
ILVBL
209L8, AHAS, ILV2H, IlvB like protein, ILVBL
Rabbit
Antigen Affinity Chromatography
RUO
10994
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A1L0T0
ILVBL
ILVBL Fusion Protein Ag21495
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers