Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10160843
  • 1470.00 SEK / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Specifications
Basket
Ultrasonic Bath, Elmasonic Xtra 30 H 3 L