Learn More
Fisherbrand™ Immersion Basket with Handle
Faciliates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11796198
  • 3908.00 SEK / Each
Estimated Shipment
07-06-2021
Add to basket
Description
Description
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Specifications
450 mm
Basket
Stainless Steel
90 L Elmasonic S 900 H Ultrasonic Unit
250 mm
545 mm