Learn More
ROBU VitraPOR™ Immersion Filter
Immersion Filter for reverse filtration.
Brand: ROBU VitraPOR™ 24354
Additional Details : Weight : 0.13200kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
4 | |
35 mm |
Non-sterile | |
210 mm |