Learn More
ROBU VitraPOR™ Immersion Filter
Immersion Filter for reverse filtration.
945.00 SEK - 1940.00 SEK
Product Code | Brand | Porosity | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
16863056
|
ROBU VitraPOR™
24204 |
4 | 20 mm | 150 mm |
1940.00 SEK
Pack of 5 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16883056
|
ROBU VitraPOR™
24351 |
1 | 35 mm | 210 mm |
945.00 SEK
Pack of 2 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16833056
|
ROBU VitraPOR™
24201 |
1 | 20 mm | 150 mm |
1830.00 SEK
Pack of 5 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16873056
|
ROBU VitraPOR™
24350 |
0 | 35 mm | 210 mm |
945.00 SEK
Pack of 2 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16853056
|
ROBU VitraPOR™
24203 |
3 | 20 mm | 150 mm |
1830.00 SEK
Pack of 5 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16813066
|
ROBU VitraPOR™
24354 |
4 | 35 mm | 210 mm |
1000.00 SEK
Pack of 2 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16843056
|
ROBU VitraPOR™
24202 |
2 | 20 mm | 150 mm |
1830.00 SEK
Pack of 5 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16823056
|
ROBU VitraPOR™
24200 |
0 | 20 mm | 150 mm |
1830.00 SEK
Pack of 5 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
16893056
|
ROBU VitraPOR™
24352 |
2 | 35 mm | 210 mm |
945.00 SEK
Pack of 2 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Non-sterile |