missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Immersion Filter

Immersion Filter for reverse filtration.

Brand:  ROBU VitraPOR™ 24200

Additional Details : Weight : 0.11970kg

 View more versions of this product

Product Code. 16823056

  • 1830.00 SEK / Pack of 5
Estimated Shipment: 06-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
5/Pk.
150 mm
Non-sterile
0
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers