missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Immersion Filter
Immersion Filter for reverse filtration.
Brand: ROBU VitraPOR™ 24201
Additional Details : Weight : 0.11970kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
1 μm | |
Non-sterile | |
150 mm |
Borosilicate Glass 3.3 | |
20 mm | |
1 |